Skip to Content
MilliporeSigma
MOPSO sodium salt BioXtra, pH 10-12 (1 M in H2O), ≥99% (titration)

MOPSO sodium salt

MOPSO sodium salt BioXtra, pH 10-12 (1 M in H2O), ≥99% (titration)
Synonym(s):
3-(N-Morpholinyl)-2-hydroxypropanesulfonic acid sodium salt, 3-Morpholino-2-hydroxypropanesulfonic acid sodium salt
Empirical Formula (Hill Notation):
C7H14NNaO5S
CAS Number:
Molecular Weight:
247.24

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Application
Showing 1-2 of 2