Skip to Content
MilliporeSigma
Monobromo(trimethylammonio)bimane bromide suitable for fluorescence, ≥90.0% (HPLC)

Monobromo(trimethylammonio)bimane bromide

Monobromo(trimethylammonio)bimane bromide suitable for fluorescence, ≥90.0% (HPLC)
Synonym(s):
(Trimethylammonio)monobromobimane, 5-(Bromomethyl)-N,N,N,2,6-pentamethyl-1,7-dioxo-1H,7H-pyrazolo[1,2-a]pyrazole-3-methanaminium bromide, Brombimane q, Thiolye MQ, qBBr
Empirical Formula (Hill Notation):
C13H19Br2N3O2
CAS Number:
Molecular Weight:
409.12

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1