Skip to Content
MilliporeSigma
Molybdenumhexacarbonyl ≥99.9% trace metals basis

Molybdenumhexacarbonyl

Molybdenumhexacarbonyl ≥99.9% trace metals basis
Synonym(s):
Hexacarbonylmolybdenum(0)
Linear Formula:
Mo(CO)6
CAS Number:
Molecular Weight:
264.00
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing