Skip to Content
MilliporeSigma
Moexipril Related Compound E pharmaceutical secondary standard, certified reference material

Moexipril Related Compound E

Moexipril Related Compound E pharmaceutical secondary standard, certified reference material
Synonym(s):
(S)-(−)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C12H15NO4 · HCl
CAS Number:
Molecular Weight:
273.71

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2