Skip to Content
MilliporeSigma
Metrafenone PESTANAL®, analytical standard

Metrafenone

Metrafenone PESTANAL®, analytical standard
Synonym(s):
(3-Bromo-6-methoxy-2-methylphenyl)(2,3,4-trimethoxy-6-methylphenyl)methanone, 3′-Bromo-2,3,4,6′-tetramethoxy-2′,6-dimethylbenzophenone
Empirical Formula (Hill Notation):
C19H21BrO5
CAS Number:
Molecular Weight:
409.27

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique