Skip to Content
MilliporeSigma
Methyl P,P-bis(2,2,2-trifluoroethyl)phosphonoacetate 95%

Methyl P,P-bis(2,2,2-trifluoroethyl)phosphonoacetate

Methyl P,P-bis(2,2,2-trifluoroethyl)phosphonoacetate 95%
Synonym(s):
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate
Linear Formula:
CH3O2CCH2P(O)(OCH2CF3)2
CAS Number:
Molecular Weight:
318.11

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing