Skip to Content
MilliporeSigma
Methylphenidate Related Compound A certified reference material, pharmaceutical secondary standard

Methylphenidate Related Compound A

Methylphenidate Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
α-Phenyl-2-piperidineacetic acid hydrochloride, Ritalinic acid hydrochloride
Empirical Formula (Hill Notation):
C13H17NO2 · HCl
CAS Number:
Molecular Weight:
255.74

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade