Skip to Content
MilliporeSigma
Methyl Caprate certified reference material, pharmaceutical secondary standard

Methyl caprate

Methyl Caprate certified reference material, pharmaceutical secondary standard
Synonym(s):
Methyl decanoate, Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate
Linear Formula:
CH3(CH2)8COOCH3
CAS Number:
Molecular Weight:
186.29

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade