Skip to Content
MilliporeSigma
Methyl 6-bromo-2-naphthoate 98%

Methyl 6-bromo-2-naphthoate

Methyl 6-bromo-2-naphthoate 98%
Linear Formula:
BrC10H6CO2CH3
CAS Number:
Molecular Weight:
265.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing