Skip to Content
MilliporeSigma
Methyl 4-aminobutyrate hydrochloride ≥99.0% (AT)

Methyl 4-aminobutyrate hydrochloride

Methyl 4-aminobutyrate hydrochloride ≥99.0% (AT)
Linear Formula:
NH2(CH2)3COOCH3 · HCl
CAS Number:
Molecular Weight:
153.61

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing