Skip to Content
MilliporeSigma
Methyl (2R)-glycidate optical purity ee: 94% (GLC), 97%

Methyl (2R)-glycidate

Methyl (2R)-glycidate optical purity ee: 94% (GLC), 97%
Synonym(s):
Methyl (R)-oxiranecarboxylate
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing