Skip to Content
MilliporeSigma
Methyl 2-methylbutyrate natural, ≥98%, FCC, FG

Methyl 2-methylbutyrate

Methyl 2-methylbutyrate natural, ≥98%, FCC, FG
Linear Formula:
CH3CH2CH(CH3)COOCH3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing