Skip to Content
MilliporeSigma
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate 97%

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate 97%
Linear Formula:
FSO2CF2CO2CH3
CAS Number:
Molecular Weight:
192.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing