Skip to Content
MilliporeSigma
meso-2,3-Dibromosuccinic acid 98%

meso-2,3-Dibromosuccinic acid

meso-2,3-Dibromosuccinic acid 98%
Linear Formula:
HO2CCH(Br)CH(Br)CO2H
CAS Number:
Molecular Weight:
275.88

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing