Skip to Content
MilliporeSigma
Medroxyprogesterone acetate Related Compound B United States Pharmacopeia (USP) Reference Standard

Medroxyprogesterone acetate Related Compound B

Medroxyprogesterone acetate Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Medroxyprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione, 17α-Hydroxy-6α-methylprogesterone, 6α-Methyl-5-pregnen-17α-ol-3,20-dione, 6-Dihydromegestrol
Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1