Skip to Content
MilliporeSigma
Manganese(II) acetylacetonate

Manganese(II) acetylacetonate

Manganese(II) acetylacetonate
Synonym(s):
2,4-Pentanedione manganese(II) derivative, Manganous acetylacetonate, Mn(acac)2, Manganese(II) acetylacetonate
Linear Formula:
[CH3COCH=C(O)CH3]2Mn
CAS Number:
Molecular Weight:
253.15

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing