Skip to Content
MilliporeSigma
Lutein Pharmaceutical Secondary Standard; Certified Reference Material

Lutein

Lutein Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Xanthophyll, α-Carotene-3,3′-diol, β, ε-carotene, β, ε-carotene-3,3′-diol, Lutein
Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3