Skip to Content
MilliporeSigma
L-Tyrosine tert-butyl ester ≥99.0% (NT)

L-Tyrosine tert-butyl ester

L-Tyrosine tert-butyl ester ≥99.0% (NT)
Linear Formula:
HOC6H4CH2CH(NH2)COOC(CH3)3
CAS Number:
Molecular Weight:
237.29
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing