Skip to Content
MilliporeSigma
Lithium L-lactate ≥98% (titration)

Lithium L-lactate

Lithium L-lactate ≥98% (titration)
Synonym(s):
(S)-2-Hydroxypropionic acid lithium salt, L-Lactic acid lithium salt, Sarcolactic acid lithium salt
Empirical Formula (Hill Notation):
C3H5LiO3
CAS Number:
Molecular Weight:
96.01
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing