Skip to Content
MilliporeSigma
Lithium bis(trimethylsilyl)amide solution 1 M in tert-butyl methyl ether

Lithium bis(trimethylsilyl)amide solution

Lithium bis(trimethylsilyl)amide solution 1 M in tert-butyl methyl ether
Synonym(s):
Hexamethyldisilazane lithium salt
Linear Formula:
[(CH3)3Si]2NLi
CAS Number:
Molecular Weight:
167.33

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Quality Segment