Skip to Content
MilliporeSigma
Lithium 2,2,6,6-tetramethylpiperidide 95%

Lithium 2,2,6,6-tetramethylpiperidide

Lithium 2,2,6,6-tetramethylpiperidide 95%
Synonym(s):
2,2,6,6-Tetramethylpiperidine lithium salt (1:1), LTMP
Empirical Formula (Hill Notation):
C9H18LiN
CAS Number:
Molecular Weight:
147.19

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing