Skip to Content
MilliporeSigma
L-Isoleucine-15N 98 atom % 15N, 98% (CP)

L-Isoleucine-15N

L-Isoleucine-15N 98 atom % 15N, 98% (CP)
Linear Formula:
CH3CH2CH(CH3)CH(15NH2)CO2H
CAS Number:
Molecular Weight:
132.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing