Skip to Content
MilliporeSigma
Lidocaine hydrochloride British Pharmacopoeia (BP) Reference Standard

Lidocaine hydrochloride

Lidocaine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Lidocaine hydrochloride monohydrate, 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate, Lignocaine hydrochloride monohydrate, Xylocaine hydrochloride monohydrate
Empirical Formula (Hill Notation):
C14H22N2O · HCl · H2O
CAS Number:
Molecular Weight:
288.81
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique