Skip to Content
MilliporeSigma
Lewis-X trisaccharide 3′-sulfate sodium salt ≥95% (TLC)

Lewis-X trisaccharide 3′-sulfate sodium salt

Lewis-X trisaccharide 3′-sulfate sodium salt ≥95% (TLC)
Synonym(s):
Lex-sulfate sodium salt, SO3(O→3)-β-Gal-(1→4)(α-Fuc-[1→3])-GlcNAc, Sulfo-Lewis-X trisaccharide sodium salt
Linear Formula:
C20H34NO18SNa
CAS Number:
Molecular Weight:
631.53

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing