Skip to Content
MilliporeSigma
L-Cystine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Cystine

L-Cystine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(R,R)-3,3′-Dithiobis(2-aminopropionic acid), L-Cystine, L-Cystine
Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique