Skip to Content
MilliporeSigma
L-Cysteinesulfinic acid monohydrate 99%

L-Cysteinesulfinic acid monohydrate

L-Cysteinesulfinic acid monohydrate 99%
Synonym(s):
3-sulfino-L-alanine
Linear Formula:
HO2SCH2CH(NH2)CO2H·H2O
CAS Number:
Molecular Weight:
171.17
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing