Skip to Content
MilliporeSigma
L-Citrulline certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Citrulline

L-Citrulline certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-2-Amino-5-ureidopentanoic acid, L-2-Amino-5-ureidovaleric acid, L-Citrulline
Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
Amino Acid
API Family
Reference Material Type
Special Grade