Skip to Content
MilliporeSigma
L-Carvone natural, 99%, FG

L-Carvone

L-Carvone natural, 99%, FG
Synonym(s):
(R)-(−)-Carvone, (−)-Carvone, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one, Carvol
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing