Skip to Content
MilliporeSigma
L-Aspartic acid sodium salt monohydrate ≥99.0% (NT)

L-Aspartic acid sodium salt monohydrate

L-Aspartic acid sodium salt monohydrate ≥99.0% (NT)
Synonym(s):
(S)-2-Aminobutanedioic acid sodium salt, Sodium L-aspartate
Linear Formula:
HO2CCH2CH(NH2)CO2Na · H2O
CAS Number:
Molecular Weight:
173.10
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2