Skip to Content
MilliporeSigma
Laquinimod ≥98% (HPLC)

Laquinimod

Laquinimod ≥98% (HPLC)
Synonym(s):
5-Chloro-N-Et-4-hydroxy-1-methyl-2-oxo-N-Ph-1,2-dihydroquinoline-3-carboxamide
Empirical Formula (Hill Notation):
C19H17ClN2O3
CAS Number:
Molecular Weight:
356.80

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing