Skip to Content
MilliporeSigma
Lamivudine British Pharmacopoeia (BP) Reference Standard

Lamivudine

Lamivudine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2′,3′-Dideoxy-3′-thiacytidine, 3TC, Epivir, Epivir-HBV, Heptovir, Zeffix, Lamivudine
Empirical Formula (Hill Notation):
C8H11N3O3S
CAS Number:
Molecular Weight:
229.26

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique