
L-Alanyl-L-alanine

Synonym(s):
Ala-Ala, L-Alanyl-L-alanine
Linear Formula:
CH3CH(NH2)CONHCH(CH3)CO2H
CAS Number:
Molecular Weight:
160.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing