Skip to Content
MilliporeSigma
Ketorolac tris salt ≥99%, crystalline

Ketorolac tris salt

Ketorolac tris salt ≥99%, crystalline
Synonym(s):
(±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol
Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40

Products

Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing