Skip to Content
MilliporeSigma
Kainic acid monohydrate ≥98% (HPLC), from Digenea simplex

Kainic acid monohydrate

Kainic acid monohydrate ≥98% (HPLC), from Digenea simplex
Synonym(s):
2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine
Empirical Formula (Hill Notation):
C10H15NO4 · H2O
CAS Number:
Molecular Weight:
231.25

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment