Skip to Content
MilliporeSigma
Isoxsuprine Hydrochloride pharmaceutical secondary standard, certified reference material

Isoxsuprine hydrochloride

Isoxsuprine Hydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
Isoxsuprine hydrochloride
Empirical Formula (Hill Notation):
C18H23NO3 · HCl
CAS Number:
Molecular Weight:
337.84

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique