Skip to Content
MilliporeSigma
Isopropyl palmitate Pharmaceutical Secondary Standard; Certified Reference Material

Isopropyl palmitate

Isopropyl palmitate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Isopropyl hexadecanoate
Linear Formula:
CH3(CH2)14COOCH(CH3)2
CAS Number:
Molecular Weight:
298.50
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique