Skip to Content
MilliporeSigma
Isopropylamine anhydrous, analytical standard

Isopropylamine

Isopropylamine anhydrous, analytical standard
Synonym(s):
2-Aminopropane, Isopropylamine
Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing