Skip to Content
MilliporeSigma
Isobutyl 1,2-dihydro-2-isobutoxy-1-quinolinecarboxylate 99%

Isobutyl 1,2-dihydro-2-isobutoxy-1-quinolinecarboxylate

Isobutyl 1,2-dihydro-2-isobutoxy-1-quinolinecarboxylate 99%
Synonym(s):
1-Isobutoxycarbonyl-2-isobutoxy-1,2-dihydroquinoline, 2-Isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline, IIDQ
Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing