Skip to Content
MilliporeSigma
Iron(III) nitrate nonahydrate ≥99.999% trace metals basis

Iron(III) nitrate nonahydrate

Iron(III) nitrate nonahydrate ≥99.999% trace metals basis
Synonym(s):
Ferric nitrate nonahydrate, Iron(III) nitrate nonahydrate
Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:

Products

Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Special Grade
Technique