Skip to Content
MilliporeSigma
Iron(III) chloride hexahydrate ≥99.99% trace metals basis

Iron(III) chloride hexahydrate

Iron(III) chloride hexahydrate ≥99.99% trace metals basis
Synonym(s):
Ferric chloride hexahydrate
Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Special Grade