Skip to Content
MilliporeSigma
Iron(0) pentacarbonyl >99.99% trace metals basis

Iron(0) pentacarbonyl

Iron(0) pentacarbonyl >99.99% trace metals basis
Synonym(s):
Iron(0) carbonyl, Pentacarbonyliron(0)
Linear Formula:
Fe(CO)5
CAS Number:
Molecular Weight:
195.90
EC Number:

Products

Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing