Skip to Content
MilliporeSigma
Insulin human ≥95% (HPLC), semisynthetic, powder, non-sterile

Insulin human

Insulin human ≥95% (HPLC), semisynthetic, powder, non-sterile
Synonym(s):
Insulin human
Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Recombinant Host
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Sterilization Method
Technique