Skip to Content
MilliporeSigma
Ibutilide hemifumarate salt ≥98% (HPLC)

Ibutilide hemifumarate salt

Ibutilide hemifumarate salt ≥98% (HPLC)
Synonym(s):
(±)-N-[4-[4-(Ethylheptylamino)-1-hydroxybutyl]phenyl]-methanesulfonamide hemifumarate salt, Corvert
Empirical Formula (Hill Notation):
C20H36N2O3S · 0.5C4H4O4
CAS Number:
Molecular Weight:
442.61

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing