Skip to Content
MilliporeSigma
Hyodeoxycholic acid ≥98%

Hyodeoxycholic acid

Hyodeoxycholic acid ≥98%
Synonym(s):
3α,6α-Dihydroxy-5β-cholan-24-oic acid
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing