Skip to Content
MilliporeSigma
Hydriodic acid distilled, 57 wt. %, 99.999% trace metals basis

Hydriodic acid

Hydriodic acid distilled, 57 wt. %, 99.999% trace metals basis
Synonym(s):
Hydriotic acid
Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade