Skip to Content
MilliporeSigma
Hexyl acetate analytical standard

Hexyl acetate

Hexyl acetate analytical standard
Synonym(s):
Capryl acetate
Linear Formula:
CH3COO(CH2)5CH3
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing