Skip to Content
MilliporeSigma
Hexane, mixture of isomers HPLC Plus, for HPLC, GC, and residue analysis, ≥98.5%

Hexane, mixture of isomers

Hexane, mixture of isomers HPLC Plus, for HPLC, GC, and residue analysis, ≥98.5%
Synonym(s):
Hexanes, Isohexane
Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Special Grade
Technique