
HBTU

Synonym(s):
N,N,N′,N′-Tetramethyl-O-(1H-benzotriazol-1-yl)uronium hexafluorophosphate, O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate, HBTU
Empirical Formula (Hill Notation):
C11H16F6N5OP
CAS Number:
Molecular Weight:
379.24
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing