![Guanidinium bromide ≥98%](/deepweb/assets/sigmaaldrich/product/structures/348/915/e2c01765-86e1-440c-a737-248d9d0b2b3d/640/e2c01765-86e1-440c-a737-248d9d0b2b3d.png)
Guanidinium bromide
![Guanidinium bromide ≥98%](/deepweb/assets/sigmaaldrich/product/structures/348/915/e2c01765-86e1-440c-a737-248d9d0b2b3d/640/e2c01765-86e1-440c-a737-248d9d0b2b3d.png)
Empirical Formula (Hill Notation):
CH6BrN3
CAS Number:
Molecular Weight:
139.98
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing