Skip to Content
MilliporeSigma
Guaiazulene phyproof® Reference Substance

Guaiazulene

Guaiazulene phyproof® Reference Substance
Synonym(s):
1,4-Dimethyl-7-isopropylazulene
Empirical Formula (Hill Notation):
C15H18
CAS Number:
Molecular Weight:
198.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing